Learn More
Spermine tetrahydrochloride, 99%, Thermo Scientific Chemicals
Binds to the polyamine modulatory site of NMDA
$60.58 - $872.71
Chemical Identifiers
CAS | 306-67-2 |
---|---|
Molecular Formula | C10H30Cl4N4 |
Molecular Weight (g/mol) | 348.18 |
MDL Number | MFCD00012914 |
InChI Key | XLDKUDAXZWHPFH-UHFFFAOYSA-N |
Synonym | spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride |
PubChem CID | 9384 |
IUPAC Name | N,N'-bis(3-aminopropyl)butane-1,4-diamine;tetrahydrochloride |
SMILES | [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6306003
|
Thermo Scientific Chemicals
J6306003 |
1 g |
N/A
|
|
|||||
AAJ6306006
|
Thermo Scientific Chemicals
J6306006 |
5 g |
N/A
|
|
|||||
AAJ6306014
|
Thermo Scientific Chemicals
J6306014 |
25 g |
N/A
|
|
|||||
Description
Spermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSpermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.
Solubility
Soluble in water (348.2 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas. Keep away from strong oxidizing agents.
Chemical Identifiers
306-67-2 | |
348.18 | |
XLDKUDAXZWHPFH-UHFFFAOYSA-N | |
9384 | |
[H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN |
C10H30Cl4N4 | |
MFCD00012914 | |
spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride | |
N,N'-bis(3-aminopropyl)butane-1,4-diamine;tetrahydrochloride |
Specifications
306-67-2 | |
C10H30Cl4N4 | |
1 g | |
spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride | |
XLDKUDAXZWHPFH-UHFFFAOYSA-N | |
N,N'-bis(3-aminopropyl)butane-1,4-diamine;tetrahydrochloride | |
9384 | |
99% | |
Spermine tetrahydrochloride |
White | |
MFCD00012914 | |
3911771 | |
Soluble in water (348.2g/L at 20°C). | |
[H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN | |
348.18 | |
348.19 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-189-8
RTECSNumber : EJ7230000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only