Please login to your online account to display your discounted pricing

Spermine NONOate, Tocris


Slow NO releasing agent

Quantity 50mg

Manufacturer: r & d systems  RDS113550

 View more versions of this product

Catalog No. 113550

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 262.35
Quantity 50mg
Assay Percent Range >98%
CAS 136587-13-8
Synonym N-[4-[1-(3-Aminopropyl)-2-hydroxy-2-nitrosohydrazino]butyl-1, 3-propanediamine
Chemical Name or Material Spermine NONOate
Molecular Formula C10H26N6O2
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Spermine NONOate
Assay Percent Range >98%
Molecular Formula C10H26N6O2
Formula Weight 262.35
Synonym N-[4-[1-(3-Aminopropyl)-2-hydroxy-2-nitrosohydrazino]butyl-1, 3-propanediamine
Accessories

Desiccate at -20°C