Please login to your online account to display your discounted pricing

Spermine NONOate, Tocris

Slow NO releasing agent

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material Spermine NONOate
Assay Percent Range >98%
Molecular Formula C10H26N6O2
Formula Weight 262.35
Synonym N-[4-[1-(3-Aminopropyl)-2-hydroxy-2-nitrosohydrazino]butyl-1, 3-propanediamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

113510

 
r & d systems
RDS113510
10mg Each for $113.57

113550

 
r & d systems
RDS113550
50mg Each for $478.51
Description & Specifications

Specifications

Formula Weight 262.35
Quantity 10mg
Chemical Name or Material Spermine NONOate
Assay Percent Range >98%
Molecular Formula C10H26N6O2
CAS 136587-13-8
Synonym N-[4-[1-(3-Aminopropyl)-2-hydroxy-2-nitrosohydrazino]butyl-1, 3-propanediamine
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C