Please login to your online account to display your discounted pricing

Spermidine Trihydrochloride, Fisher BioReagents


Quantity 25g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP254025

 View more versions of this product

Catalog No. BP254025

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 254.63
Melting Point 248°C
Quantity 25g
Physical Form Solid
Molecular Formula C7H19N3 · 3 HCl
Color White
pH 3 to 6
Merck Index 15, 8869
Chemical Name or Material Spermidine Trihydrochloride
CAS 334-50-9
Packaging Amber Glass
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Irritating to eyes, respiratory system and skin. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:1
Recommended Storage Store below 20°C, desiccate.
FTIR Conforms to reference
Identification Pass Test
Assay Percent Range ≥99 %
ChemAlert Storage Symbol Gray
Purity 99% min.

Specifications

Chemical Name or Material Spermidine Trihydrochloride
CAS 334-50-9
Physical Form Solid
Assay Percent Range ≥99 %
Molecular Formula C7H19N3 · 3 HCl
Formula Weight 254.63

  • Spermidine is an aliphatic polyamine used in various biochemical applications due to its involvement in many metabolic functions within organisms
  • Forms stable compounds with nucleic acids and may be used for the purification of DNA-binding proteins
  • Also serves as a plant growth regulator

Accessories
 

Emergency Overview
Irritating to eyes, respiratory system and skin. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:1

WARNING!

Store below 20°C, desiccate.

SDS & Safety Documents