Learn More
Spermidine Trihydrochloride, Fisher BioReagents
$142.45 - $338.60

Chemical Identifiers
CAS | 334-50-9 |
---|---|
Molecular Formula | C7H22Cl3N3 |
Molecular Weight (g/mol) | 254.62 |
MDL Number | MFCD00012918 |
InChI Key | LCNBIHVSOPXFMR-UHFFFAOYSA-N |
Synonym | 1,4-butanediamine, n-3-aminopropyl , hydrochloride, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, 4-azoniaoctamethylenediammonium trichloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, spermidine hydrochloride, spermidine trihydrochloride, spermidine, trihydrochloride, unii-1o14bed398 |
PubChem CID | 9539 |
IUPAC Name | N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride |
SMILES | [H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP25401
|
Fisher BioReagents
BP25401 |
1g | Amber Glass |
Each for $142.45
Case of 9 Each for $1,282.05 |
|
BP25405
|
Fisher BioReagents
BP25405 |
5g | Amber Glass |
Each for $338.60
Case of 9 Each for $3,047.40 |
|
Description
Spermidine is an aliphatic polyamine used in various biochemical applications due to its involvement in many metabolic functions within organisms
Forms stable compounds with nucleic acids and may be used for the purification of DNA-binding proteins
Chemical Identifiers
334-50-9 | |
254.62 | |
LCNBIHVSOPXFMR-UHFFFAOYSA-N | |
9539 | |
[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN |
C7H22Cl3N3 | |
MFCD00012918 | |
1,4-butanediamine, n-3-aminopropyl , hydrochloride, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, 4-azoniaoctamethylenediammonium trichloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, spermidine hydrochloride, spermidine trihydrochloride, spermidine, trihydrochloride, unii-1o14bed398 | |
N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride |
Specifications
334-50-9 | |
White | |
Pass Test | |
C7H22Cl3N3 | |
Amber Glass | |
1,4-butanediamine, n-3-aminopropyl , hydrochloride, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, 4-azoniaoctamethylenediammonium trichloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, spermidine hydrochloride, spermidine trihydrochloride, spermidine, trihydrochloride, unii-1o14bed398 | |
LCNBIHVSOPXFMR-UHFFFAOYSA-N | |
N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride | |
9539 | |
Solid | |
Spermidine Trihydrochloride |
3 to 6 | |
248°C | |
≥99 % | |
MFCD00012918 | |
15, 8869 | |
1g | |
[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN | |
254.62 | |
254.63 | |
99% min. |
Safety and Handling

WARNING!
Emergency Overview
Irritating to eyes, respiratory system and skin. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:2
Flammability:1
Instability:1
Recommended Storage : Store below 20°C, desiccate.