Please login to your online account to display your discounted pricing

Spermidine, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC132740250

 View more versions of this product

Catalog No. AC132740250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 145.25
Physical Form Liquid After Melting
Quantity 25g
Color Clear colorless
Density 0.925g/mL
MDL Number MFCD00008229
Assay Percent Range 99%
Molecular Formula C7H19N3
Health Hazard 1 GHS Signal Word: Danger
Refractive Index 1.4785 to 1.4805 (20°C, 589nm)
Packaging Glass bottle
Synonym N-(3-Aminopropyl)-1, 4-butanediamine
Infrared Spectrum Authentic
Chemical Name or Material Spermidine
CAS 124-20-9
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Spermidine
CAS 124-20-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H19N3
Formula Weight 145.25
MDL Number MFCD00008229
Synonym N-(3-Aminopropyl)-1, 4-butanediamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents