missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ SAF-T-FIT™ Plus N95 Particulate Respirators

For use with particulate aerosols free of oil

Manufacturer:  Honeywell™ 14110388

 View more versions of this product

Catalog No. 29-635-5341




Description

Description

  • Low breathing resistance
  • Incorporates humidity-resistant filter media
  • Durable suspension straps
  • Packaged to maintain integrity of product in work environment
  • Adjustable boomerang nose seal for secure fit
  • Color coded nose clips
  • Latex-free

Specifications

Specifications

N - Not Resistant to Oil
White
Yes
General Purpose Particulate Respirator
No
Particulate aerosols free of oil, fire protection, medical, pharmaceutical, transportation
14110388
Two-Strap Design
NIOSH 42 CFR 84, N95, NIOSH approval number: TC-84A-3789
Cup/Molded
N95
Medium
95%
Yes
20/Pk.
No
SDS
Documents

Documents

Product Certifications
Promotions

Promotions