missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ North™ SAF-T-FIT™ Plus N95 Particulate Respirators

For use with particulate aerosols free of oil

Manufacturer:  Honeywell™ 14110399

 View more versions of this product

Catalog No. 29-635-5352




Description

Description

  • Low breathing resistance
  • Incorporates humidity-resistant filter media
  • Durable suspension straps
  • Packaged to maintain integrity of product in work environment
  • Adjustable boomerang nose seal for secure fit
  • Color coded nose clips
  • Latex-free

Compliance

Meets NIOSH TC-84A-3789; with filter efficiency level of at least 95% against particulates and non-petroleum based liquid aerosols.

Specifications

Specifications

Yes
NIOSH TC-84A-3789
Gray
Small
Welding, soldering, brazing, torch cutting, metal pouring, grinding, sanding, particulate aerosols free of oil
Yes
Humidity
N95 Filter
Red
N1125Ag
95% against solid particulates and non-petroleum based liquid aerosols
Boomerang nose seal and valve
10/Pk.
SDS
Documents