Honeywell™ North™ SAF-T-FIT™ N99 Disposable Particulate Respirator
N99 protection combined with a full sealing flange for superior fit and protection
Manufacturer: Honeywell™ 14110402
Description
- Adjustable straps for a snug fit
- Exhalation valve for ease of breathing during extended wear
- Boomerang nose seal is adjustable for added comfort and a secure fit
- Nose clip is color coded for visible size recognition
- NIOSH Approval TC-84A-3845
- Latex-free
Compliance
Meets NIOSH TC-84A-3845; filter efficiency level of at least 99% against solid particulates and non-petroleum based liquied aerosols.
Specifications
Yes | |
NIOSH TC-84A-3845 | |
Particulate Respirator | |
Welding, soldering, metal cutting and pouring, torch cutting, grinding, sanding, sweeping, hot temperatures, general industrial applications and dusty operation | |
10/Pk. |
N99 Filter | |
White | |
Small | |
Yes |