Sperian Premier Plus Half-Mask Respirators
Manufacturer: North Safety Products 321000
Description
- Unique design offers improved comfort
- Unlike traditional pear-shaped masks, respirator has a wraparound style that positions the cartridges farther back on the head, widening the field of vision
- Floating yoke design enables adjustment of the headband without disturbing the fit of the respirator
- Triple-sealing flange provides additional maneuverability, resulting in a more secure fit on a wider range of faces
- Nonslip cradle suspension and soft nosepiece leaves ample room for protective eyewear
- Utilizes Sperian S-series cartridges and filters and also accommodates the multi-contaminant cartridge for use in multiple contaminant environments
Specifications
Silicone | |
Small | |
Triple sealing flange |
North™ Premier Plus Half Mask Respirator | |
S series cartridge |