Please login to your online account to display your discounted pricing

Sperian One-Fit Healthcare N95 Particulate Respirator and Surgical Mask

Fit most face shapes and sizes while offering the ultimate in protection and comfort for healthcare settings

$22.96 - $22.99

Specifications

Certifications NIOSH, CDC
Color White
Disposable Yes
Filter Type N95 Filter
For Use With (Application) Designed for healthcare settings and use in emergency rooms, operating rooms and intensive care units
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

29-636-0703

 
north safety products
14110446H5
Molded Cup Pack of 20 Each for $22.96
Case of 10 Pack for $219.50

29-636-0759

 
north safety products
14110451
Flat form Pack of 20 Each for $22.99
Description & Specifications

Specifications

Certifications NIOSH, CDC
Color White
Disposable Yes
Filter Type N95 Filter
For Use With (Application) Designed for healthcare settings and use in emergency rooms, operating rooms and intensive care units
Quantity 20
Size Universal

Designed for healthcare settings and use in Emergency Rooms, Operating Rooms and Intensive Care Units.

  • NIOSH-approved as an N95 Particulate Respirator
  • Meets CDC guidelines for M Tuberculosis exposure control
  • Ranks as “High Barrier” class 160mmHg in conformance to ASTM F1862 requirements for fluid penetration and splash resistance
  • Reduces the transfer of microorganisms and other airborne particulate matter
  • Reduces wearer exposure to airborne particulates smaller than 0.1 micron—appropriate for laser and electrocautery procedures

  • >99.9% (BFE) Bacterial Filtration Efficiency
  • >99.5% (PFE) Particle Filtration Efficiency
  • >99.9% (VFE) Viral Filtration Efficiency