Honeywell™ Sperian™ Fit-Test Kits
Sperian offers various Qualitative (QLFT) fit test kits for all your testing needs.
$156.62 - $156.62
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
17-648-3
|
Honeywell™
140090 |
Irritant Smoke Fit Test Kit |
Each for $156.62
|
|
Description
Two kits with all the hard-to-find items needed to perform the OSHA accepted qualitative fit tests for irritant smoke and banana oil. Include step-by-step procedures, test exercises, and a reproducible “Respirator Fit-Test Record” form for help in complying with OSHA regulations.
Specifications
OSHA | |
1 Ea. |