Sper Scientific™ RH/Temperature SD Card Logger
Reads relative humidity and ambient temperature dew point and wet bulb, and functions as a type K or J thermometer.
Manufacturer:  Fisher Scientific 800021C
 View more versions of this product
Catalog No. S05860C
Specifications Description & Specifications
Specifications
454g
0.001
Rh/Temperature Sd Card Logger, Certified
Reads relative humidity and ambient temperature dew point and wet bulb and functions as a type K or J thermometer
LCD
5 to 95%
-32° to 122°F
1 lb.
CE, RoHS
Detachable probe, six AA batteries, fold-out tripod, instruction manual, a 2GB SD card and a soft carrying case
0° to 50°C
±3%