Sper Scientific™ RH/Temperature SD Card Logger
Reads relative humidity and ambient temperature dew point and wet bulb, and functions as a type K or J thermometer.
Manufacturer:  Fisher Science Education 800021
 View more versions of this product
Catalog No. S05860
Specifications Description & Specifications
Specifications
0.001
±3%
CE, RoHS
1 lb.
-32° to 122°F
Rh/Temperature Sd Card Logger
Reads relative humidity and ambient temperature dew point and wet bulb and functions as a type K or J thermometer
LCD
0° to 50°C
454g
Detachable probe, six AA batteries, fold-out tripod, instruction manual, a 2GB SD card and a soft carrying case
5 to 95%