Sper Scientific™ Pocket Digital Refractometers
Measure brix, salinity and many other parameters in the field with laboratory-level accuracy.
Manufacturer:  Fisher Science Education 300054
 View more versions of this product
Catalog No. S96271
Specifications Description & Specifications
Specifications
±0.2%
LCD
32mm
4.63 in.
118mm
130g
62mm
0 to 28%
1.25 in.
Distilled Water, transfer pipet, AAA battery, instructions and case
0.1%
4.6 oz.
2.44 in.
4.63 x 2.44 x 1.25 in. (118 x 62 x 32mm)