Please login to your online account to display your discounted pricing

Sper Scientific™ pH SD Card Logger


Reads and records pH, ORP and temperature data onto standard SD memory cards.

Manufacturer: fisher scientific  850060

 View more versions of this product

Catalog No. S05864

  • / Each
 

Description & Specifications

Specifications

Warranty 1 year
Display Type LCD
Includes One pH SD Card Logger, fold-out tripod, soft carrying case, six AA batteries, instruction manual and one 2GB SD card
Range (mV) -1999 to 1999mV
Resolution (mV) 1mV
Certifications/Compliance CE, RoHS
Battery Type AA
For Use With (Application) pH, ORP, Temperature
Detection Range 0 to 14pH
Type pH SD Card Logger
Range (pH) 2 to 12
Temperature (Metric) Operating 0 to 50°C
Accuracy (pH) ±0.02
Accuracy (mV) ±0.5% +2mV
Resolution (pH) 0.01