Please login to your online account to display your discounted pricing

Sper Scientific™ pH SD Card Logger


Reads and records pH, ORP and temperature data onto standard SD memory cards.

Manufacturer: fisher science education  850061

 View more versions of this product

Catalog No. S05865

  • / Each
 

Description & Specifications

Specifications

Includes One pH SD Card Logger, sealed Epoxy body reference combination electrode with a BNC connector and a 2.5 ft cable, ATC temperature probe, pH buffers 4, 7, 10 and deionized water, 6 AA batteries and instructions; hard-shell, foam-lined case.
Range (pH) 2 to 12
Resolution (pH) 0.01
Accuracy (pH) ±0.02
Range (mV) -1999 to 1999mV
Resolution (mV) 1mV
Accuracy (mV) ±0.5% +2mV
Display Type LCD
Outputs RS232/USB PC computer interface
Battery Type AA
Certifications/Compliance CE, RoHS
Temperature (English) Operating 32° to 122°F
Temperature (Metric) Operating 0 to 50°C
For Use With (Application) pH, ORP, Temperature
Type pH SD Card Logger Kit
Warranty 1 year