Sper Scientific™ Mini Environmental Quality Meters
Monitor up to 12 environmental parameters with one tiny instrument
Manufacturer:  Fisher Science Education 850027
 View more versions of this product
Catalog No. S07073
Specifications Description & Specifications
Specifications
CE
Monitors Wind Speed, Air Temperature, Air Volume, Relative Humidity, Wind Chill, Heat Stress Index, Dew Point, Wet Bulb, PT 1000 ohm Probe, Baromatric Pressure, Altitutde, Visible Light and Compass
5 sec.
12-Paramaters Mini Environmental Quality Meter
160g (battery included)
Wrist strap, carry pouch, battery and instructions