Please login to your online account to display your discounted pricing
Sper Scientific™ Mini Environmental Quality Meters
Monitor up to 12 environmental parameters with one tiny instrument
Manufacturer: fisher science education  850026
 View more versions of this product
Catalog No. S07072
  • / Each
Description & Specifications
Specifications
For Use With (Application) Monitors Wind Speed, Air Temperature, Air Volume, Relative Humidity, Wind Chill, Heat Stress Index, Dew Point and Wet Bulb
Includes Wrist strap, carry pouch, battery and instructions
Test Time 5 sec.
Weight (Metric) 160g (battery included)
Product Type 7-Paramaters Mini Environmental Quality Meter
Certifications/Compliance CE