Please login to your online account to display your discounted pricing
Sper Scientific™ Mini Environmental Quality Meters
Monitor up to 12 environmental parameters with one tiny instrument
Manufacturer: fisher science education  850025
 View more versions of this product
Catalog No. S07070
  • / Each
Description & Specifications
Specifications
For Use With (Application) Monitors Wind Speed, Air Temperature and Relative Humidity
Includes Wrist strap, carry pouch, battery and instructions
Test Time 5 sec.
Weight (Metric) 160g (battery included)
Certifications/Compliance CE
Product Type 3-Paramaters Mini Environmental Quality Meter