Please login to your online account to display your discounted pricing
Sper Scientific™ Laser Pointers
Ideal teaching aids for classroom use and more.
$59.75 - $129.60
Products
Catalog Number Mfr. No. Product Type Price Quantity    
S68611
fisher science education
330001
630-680nm Laser Pointers Each for $129.60
S68612
fisher science education
330006
UV/Laser/White Light Pointer Each for $59.75
Description & Specifications
Laser Pointer
  • Sturdy brushed-metal pen-shaped unit is easily carried in a shirt pocket
  • Projects a brilliant, red dot visible even in bright, ambient light or with video, film and slide presentations
  • Runs for a minimum of four continuous hours on two standard alkaline batteries (included)
  • Use as a teaching tool for science presentations, conferences, lectures, seminars and planetariums
UV/Laser/White Light Pointer
  • Pocket-sized unit on key chain device
  • Press one end of the button to get a bright 650nm red laser pointer
  • Press the other end and get high-intensity white LED light for inspection and illumination
  • Press the same side again and get 400nm ultraviolet light
  • Four button cell batteries included
CE listed
Five years