missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sper Scientific™ IR Thermometers

Serve a wide variety of temperature monitoring functions.

$91.30 - $106.75

Specifications

Accuracy ± 2°C
Certifications/Compliance CE, UL
Display LCD
Length (English) Overall 8 in.
Length (Metric) Overall 203mm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Resolution Temperature Range (English) Temperature Range (Metric) Price Quantity  
Catalog Number Mfr. No. Resolution Temperature Range (English) Temperature Range (Metric) Price Quantity  
S98763 Fisher Science Education
800102
0.2°C -4° to +930°F -20° to +500°C Each for $106.75
S98762 Fisher Science Education
800101
0.5°C -4° to +605°F -20° to +320°C Each for $91.30
Specifications

Description

Safely measure the temperature of hazardous objects in electrical, hazmat situations; HVAC applications; moving machinery in factories, automobiles; electrical troubleshooting. Avoid contamination in the food, beverage and pharmaceutical industries.

  • Hold IR gun, press trigger and aim laser guide at target
  • Results appear in °C or °F on backlit LCD and are automatically held for six seconds after trigger is released
  • 8:1 distance to spot ratio (D:S)
  • Measure up to 500°C (930°F) with 0.1° resolution
  • Auto power off
  • Emissivity: 0.95 fixed
  • Range: -20 to +500°C; -4 to +930°F
  • Weight: 6 oz. (170g)
  • Dimensions: 6.125 × 3.375 × 1.25 in. (15.6 × 8.7 × 3.2cm)
Model 800101
  • Accuracy: ±2% of reading or ±2°C, whichever is greater
  • Resolution 0.5°
Model 800102
  • Accuracy: ±2°C (±3°F) from -20 to +100°C (-4 to +212°F), ±2% from 100 to 500°C (212 to 930°F)
  • Resolution 0.2°

Warranty

Five-year

Description & Specifications

Specifications

± 2°C
LCD
203mm
0.17kg
CE, UL
8 in.
6 oz.