Please login to your online account to display your discounted pricing
Sper Scientific Environmental Quality Meter with Sound
Measures air speed, humidity, light, temperature and sound.
Manufacturer: fisher science education  850069C
 View more versions of this product
Catalog No. S99077
  • / Each
Description & Specifications
Specifications
For Use With (Application) Measures air speed, humidity, light, temperature and sound
Product Type Traceable™ Certificate
For Use With (Equipment) Environmental quality meter