Please login to your online account to display your discounted pricing
Sper Scientific Environmental Quality Meter with Sound
Measures air speed, humidity, light, temperature and sound.
$407.25 - $456.75
Products
Catalog Number Mfr. No. Price Quantity    
S99077
fisher science education
850069C
Each for $456.75
S99076
fisher science education
850069
Each for $407.25
Description & Specifications
Combines an anemometer, hygrometer, thermometer, sound and light meter into a single handheld unit.
  • Reads any type-K thermocouple probe
  • results are displayed on a large dual LCD with low battery and out of range indications
  • Features touch-tone buttons, min/max, auto power off and hold functions
  • Soft carrying case with RS-232 computer port, instructions, 6 ”AAA” batteries, tripod back and a tripod screw
«
Five years