Please login to your online account to display your discounted pricing
Sper Scientific™ Benchtop Water Quality Meters
Ruggedly housed, large LCD displays the parameter with time, date and temperature.
$301.05 - $301.05
Specifications
Certifications/Compliance CE
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
S98755
fisher scientific
860031
Benchtop pH/MV Meter Each for $301.05
Description & Specifications
Specifications
Certifications/Compliance CE
  • Auto or manual temperature compensation and ranging
  • Auto buffer recognition; digital/analog outputs
  • Min./max. and 99 data points; data hold
  • Electrode status indication; operates on AC voltage
  • Come ready to use with AC adapter, computer cable and software
  • L × W × H: 8.5 × 7 × 2.25 in. (21.7 × 16.8 × 5.8cm)
pH/mV Meter
  • Five calibration points
  • Includes ATC pH electrode and electrode holder arm
  • Accepts any pH or ORP probe with a standard BNC connector
Conductivity/TDS/Salinity Meter
  • Five calibration points of Conductivity and TDS, two for Salinity
  • Includes ATC conductivity electrode and electrode holder arm
Combination Water Quality Meter
  • Reads pH, ORP, MV, Conductivity, TDS, Salinity and Temperature
  • Five calibration points
  • Includes ATC conductivity and pH electrodes and multiple electrodes holder arm
  • Accepts any pH or ORP probe with a standard BNC connector