missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing
Selected MWCOs from 100 to 100,000 Daltons
Supplier: Spectrum™ 131270
Description
Selected MWCOs from 100 to 100,000 Daltons
- CE tubing membranes are ideal for desalting, purification, and molecular separation
- Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution
Order Info
Note:Do not tie—must be used with Spectra/Por nylon closures (08-671-52, -53, -55). Strong polar solvents such as acetone, MEK, or dioxane will irreparably damage membranes. Contact Spectrum Technical Service, 1-800-634-3300, for more information.
Specifications
| Spectra/Por Dialysis Membrane Tubing | |
| Cellulose Ester | |
| 0.39 in. | |
| 10 m | |
| 39.2°F to 98.6°F | |
| Each |
| 10 kDa | |
| 10 mm | |
| 33 ft. | |
| Supplied in 0.1% sodium azide solution | |
| 4°C to 37°C | |
| Ideal for desalting, purification, and molecular separation |