Please login to your online account to display your discounted pricing

Spectrum™ Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO


Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

Manufacturer: spectrum™  SML132645

 View more versions of this product

Catalog No. 08-670-3

  • / Each
 

Description & Specifications

Specifications

Product Type Spectra/Por™ 1 Dialysis Tubing
Length (Metric) 15.2m
Temperature Range (English) 39.2° to 249.8°F
Diameter (English) 0.25 in.
Temperature Range (Metric) 4° to 121°C
Diameter (Metric) 6.4mm
MWCO 6000 to 8000 d
Quantity Each
Formulation Contain glycerin as a humectant
Material Regenerated Cellulose
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Length (English) 49.8 ft.

Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.

  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus

Do not tie knots in tubing; use closures