Please login to your online account to display your discounted pricing

Spectrum™ Clear Bath

Keeps water baths, incubators, temperature controlled instruments, humidifiers, etc. clean and clear

$125.03 - $217.94

Specifications

Boiling Point 200°F
Certifications/Compliance US OSHA, CFR 1910.1200, WHMIS
Color Pale Yellow
Composition Benzalkonium Chloride, n-alkyl (50% C14, 40% C12, 10% C16) Dimethyl Benzyl Ammonium Chloride as active ingredients, Water and Ethyl Alcohol as inert ingredients
For Use With (Application) Used to keep any circulating, temperature-controlled instrument clean, instruments like circulating Water baths, incubators, temperature controlled instruments, humidifiers, automatic analyzer baths and Blood/gas analyzers
View More Specs
Products
Catalog Number Mfr. No. Volume (English) Volume (Metric) Price Quantity    

08-671-200

 
spectrum™
SML105535
2 oz. 57.5mL Each for $125.03

11-190-316

 
spectrum™
SML105540
8 oz. 230mL Each for $217.94
Description & Specifications

Specifications

Boiling Point 200°F
Certifications/Compliance US OSHA, CFR 1910.1200, WHMIS
Color Pale Yellow
Composition Benzalkonium Chloride, n-alkyl (50% C14, 40% C12, 10% C16) Dimethyl Benzyl Ammonium Chloride as active ingredients, Water and Ethyl Alcohol as inert ingredients
For Use With (Application) Used to keep any circulating, temperature-controlled instrument clean, instruments like circulating Water baths, incubators, temperature controlled instruments, humidifiers, automatic analyzer baths and Blood/gas analyzers
Format Liquid
pH Range 6.5 to 8.3

  • Does not contain copper or other heavy metals