missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Spectrum™ MediaKap™ and MediaKap Plus HF Filters

0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling

Manufacturer:  Spectrum™ MP2M05018S

 View more versions of this product

Catalog No. 08-671-4C


 

Specifications

Description

Bottle Top Filter Unit; MediaKap-5 Plus; Capacity: 2 to 5L; w/out Fill Bell; Filtration Area: 70cm2; Pore Size: 0.2um; Sterile; Input/Output Connections: Female Luer Lok/Male Luer Lok; Mixed cellulose ester membrane

  • Efficiently sterilize and clarify culture medium or buffer solution
  • Smaller, narrow design for low hold-up volume
  • Polypropylene vent prevents air lock
  • Operates with peristaltic pump, vacuum, or syringe
  • Available in five different sizes that filter designated volume in 15 to 20 minutes
  • Sterile, biocompatible and nonpyrogenic
  • USP Class VI; 100% integrity tested

  • Optional filling bell maintains sterility while filling bottles
  • MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium

Description & Specifications

Specifications

Yes
18 Case
Female luer lok/0.25HB
MediaKap-5 Plus
750mL/min. at 10psi
Automatic vent to prevent air lock
2 to 5L
0.2μm
Polycarbonate, Polyurethane
Bottle Top Filter Unit
70cm2
Efficiently sterilize and clarify culture medium or buffer solution
Mixed Cellulose Ester