Please login to your online account to display your discounted pricing

Spectrum™ MediaKap™ and MediaKap Plus HF Filters


0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling

Manufacturer: spectrum™  MP2M50B03S

 View more versions of this product

Catalog No. 08-671-4G

  • / Pack of 3 Each
 

Description & Specifications

Specifications

Sterile Yes
Pore Size (Metric) 0.2μm
Quantity (No. per Case) 3 Case
Material Polycarbonate, Polyurethane
Connectors VarHB/VarHB
Product Type Bottle Top Filter Unit
Filter Type MediaKap-50 Plus
Filtration Area 440cm2
Flow Rate 2000mL/min. at 10psig
For Use With (Application) Efficiently sterilize and clarify culture medium or buffer solution
Includes Automatic vent to prevent air lock, filling bell
Material (Membrane) Mixed Cellulose Ester
Volume (Metric) 25 to 50L

  • Efficiently sterilize and clarify culture medium or buffer solution
  • Smaller, narrow design for low hold-up volume
  • Polypropylene vent prevents air lock
  • Operates with peristaltic pump, vacuum, or syringe
  • Available in five different sizes that filter designated volume in 15 to 20 minutes
  • Sterile, biocompatible and nonpyrogenic
  • USP Class VI; 100% integrity tested

  • Optional filling bell maintains sterility while filling bottles
  • MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium