Please login to your online account to display your discounted pricing

Spectrum™ MediaKap™ and MediaKap Plus HF Filters


0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling

Manufacturer: spectrum™  ME2M50B03S

 View more versions of this product

Catalog No. 08-671-2H

  • / Pack of 3 Each
 

Description & Specifications

Specifications

Product Type Bottle Top Filter Unit
Filter Type MediaKap-50
Volume (Metric) 25 to 50L
Pore Size (Metric) 0.2μm
Quantity (No. per Case) 3 Case
Flow Rate 2000mL/min. at 10psig
Material Polycarbonate, Polyurethane
Filtration Area 440cm2
Includes Automatic vent to prevent air lock, filling bell
Connectors VarHB/VarHB
For Use With (Application) Efficiently sterilize and clarify culture medium or buffer solution
Material (Membrane) Mixed Cellulose Ester
Sterile Yes

  • Efficiently sterilize and clarify culture medium or buffer solution
  • Smaller, narrow design for low hold-up volume
  • Polypropylene vent prevents air lock
  • Operates with peristaltic pump, vacuum, or syringe
  • Available in five different sizes that filter designated volume in 15 to 20 minutes
  • Sterile, biocompatible and nonpyrogenic
  • USP Class VI; 100% integrity tested

  • Optional filling bell maintains sterility while filling bottles
  • MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium