Please login to your online account to display your discounted pricing

Spectrum™ MediaKap™ and MediaKap Plus HF Filters


0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling

Manufacturer: spectrum™  ME2M25B06S

 View more versions of this product

Catalog No. 08-671-2G

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Product Type Bottle Top Filter Unit
Filter Type MediaKap-25
Filtration Area 185cm2
For Use With (Application) Efficiently sterilize and clarify culture medium or buffer solution
Includes Automatic vent to prevent air lock, filling bell
Sterile Yes
Volume (Metric) 10 to 25L
Pore Size (Metric) 0.2μm
Quantity (No. per Case) 6 Case
Material Polycarbonate, Polyurethane
Connectors VarHB/VarHB
Flow Rate 1400mL/min. at 10psi
Material (Membrane) Mixed Cellulose Ester