Please login to your online account to display your discounted pricing

Spectrum™ MediaKap™ and MediaKap Plus HF Filters


0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling

Manufacturer: spectrum™  ME2M10B12S

 View more versions of this product

Catalog No. 08-671-2F

  • / Pack of 12 Each
 

Description & Specifications

Specifications

Filtration Area 100cm2
Material Polycarbonate, Polyurethane
Volume (Metric) 5 to 10L
Product Type Bottle Top Filter Unit
Filter Type MediaKap-10
Flow Rate 1000mL/min. at 10psig
Pore Size (Metric) 0.2μm
Quantity (No. per Case) 12 Case
Includes Automatic vent to prevent air lock, filling bell
Connectors 0.25HB/0.25HB
Material (Membrane) Mixed Cellulose Ester
For Use With (Application) Efficiently sterilize and clarify culture medium or buffer solution
Sterile Yes

  • Efficiently sterilize and clarify culture medium or buffer solution
  • Smaller, narrow design for low hold-up volume
  • Polypropylene vent prevents air lock
  • Operates with peristaltic pump, vacuum, or syringe
  • Available in five different sizes that filter designated volume in 15 to 20 minutes
  • Sterile, biocompatible and nonpyrogenic
  • USP Class VI; 100% integrity tested

  • Optional filling bell maintains sterility while filling bottles
  • MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium