Please login to your online account to display your discounted pricing

Spectrum™ MediaKap™ and MediaKap Plus HF Filters


0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling

Manufacturer: spectrum™  ME2M05B12S

 View more versions of this product

Catalog No. 08-671-2D

  • / Pack of 12 Each
 

Description & Specifications

Specifications

Material Polycarbonate, Polyurethane
Filter Type MediaKap-5
Includes Automatic vent to prevent air lock, filling bell
Pore Size (Metric) 0.2μm
Quantity (No. per Case) 12 Case
Material (Membrane) Mixed Cellulose Ester
Product Type Bottle Top Filter Unit
For Use With (Application) Efficiently sterilize and clarify culture medium or buffer solution
Connectors Female luer lok/male luer slip
Flow Rate 750mL/min. at 10psi
Volume (Metric) 2 to 5L
Filtration Area 70cm2
Sterile Yes