Spectrum™ Labs Spectra/Por™ 7 25,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
Manufacturer:  Spectrum™ SML132125T
 View more versions of this product
Catalog No. 08-700-219
Includes:
  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks
Specifications
Description
Kit, Dialysis; Spectrum Labs; Spectra/Por 7; Standard RC; Pre-treated; Includes 1m pre-treated tubing, a pair of closures, 5 opening picks; 1m tubing/kit; 25,000 D MWCO; 12mm width; 7.5mm dia.; Vol./Length: 0.45mL/cm
  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application
Description & Specifications
Specifications
0.45mL/cm vol./Length
7.5mm
1m tubing/kit
25 kD