Please login to your online account to display your discounted pricing

Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits


With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

Manufacturer: spectrum™  SML132724T1

 View more versions of this product

  • 1m or 5m of Spectra/Por 3 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers

Catalog No. 08-700-164

  • / Each
 

Description & Specifications

Specifications

MWCO 3.5 kD
Diameter (Metric) 29mm
Quantity 1m tubing/kit
Length (Metric) 6.4mL/cm vol./Length

  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)