Please login to your online account to display your discounted pricing

Spectrum™ Labs Spectra/Por™ 2 12-14 kD MWCO Standard RC Dry Dialysis Kits


With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

Manufacturer: spectrum™  SML132678T1

 View more versions of this product

  • 1m or 5m of Spectra/Por 2 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers

Catalog No. 08-700-152

  • / Each
 

Description & Specifications

Specifications

Length (Metric) 2mL/cm
Diameter (Metric) 16mm
Quantity 1m tubing/kit
MWCO 12-14 kD

  • For higher permeability applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)