Please login to your online account to display your discounted pricing

Spectrum™ Trial Size Kits for Standard-Grade RC Dialysis Membrane Tubing (5 meter)

Ideal for smaller basic dialysis applications

$317.24 - $354.88

Specifications

Autoclavable Yes
Membrane Type Regenerated Cellulose
Product Type Trial Kit
Quantity Each
For Use With (Application) Ideal for small volume dialysis or evaluation of membrane for an application
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Diameter (English) Diameter (Metric) MWCO Price Quantity    

08-801-241

 
spectrum™
SML132655T
3.3mL 0.8 in. 20.4mm 6 to 8 kd Each for $337.85

08-801-243

 
spectrum™
SML132678T
2mL 0.6 in. 16mm 12 to 14 kd Each for $317.24

08-801-245

 
spectrum™
SML132720T
1.1mL 0.4 in. 11.5mm 3.5 kd Each for $337.85

08-801-242

 
spectrum™
SML132665T
7.9mL 1.2 in. 32mm 6 to 8 kd Each for $353.24

08-801-244

 
spectrum™
SML132680T
6.4mL 1.1 in. 29mm 12 to 14 kd Each for $353.24

08-801-246

 
spectrum™
SML132725T
9.3mL 0.2 in. 34mm 3.5 kd Each for $354.88
Description & Specifications

Specifications

Autoclavable Yes
Membrane Type Regenerated Cellulose
Product Type Trial Kit
Quantity Each
For Use With (Application) Ideal for small volume dialysis or evaluation of membrane for an application
Kit Contents 5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing, 1 standard spectra/por closure (aids buoyancy), 1 weighted spectra/por closure (aids vertical orientation), 5 opening picks
Length (English) 16 ft.
Length (Metric) 5m
Shelf Life 2 Years

Multi-purpose standard Regenerated Cellulose (RC) tubing is now available in 5m length trial kits with a pair of tubing closures. Uses include desalting, pH Change or buffer exchange or evaluation of membrane for an application.

Kit Includes:

  • 5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing
  • 1 standard Spectra/Por Closure (aids buoyancy)
  • 1 weighted Spectra/Por Closure (aids vertical orientation)
  • 5 opening picks

Safety and Handling

2 Years