Please login to your online account to display your discounted pricing
Spectrum™ Trial Size Kits for Biotech-Grade RC Dialysis Membrane Tubing
0.5 meter (1.6 ft) dialysis tubing length ideal for small applications or membrane evaluation
Manufacturer: spectrum™  SML133270T
 View more versions of this product
Catalog No. 08-607-070
  • / Each
Description & Specifications
Specifications
Product Type Dialysis Trial Kit
Quantity Each
Kit Contents 0.5m Biotech RC tubing with glycerine, 2 universal tubing closures, 5 opening picks
MWCO 8 to 10 kd
Diameter (English) 0.39 in.
For Use With (Application) Ideal for membrane and application evaluation for more critical separations
Diameter (Metric) 10mm