Please login to your online account to display your discounted pricing

Spectrum™ Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO


Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane

Manufacturer: spectrum™  SML132625

 View more versions of this product

Catalog No. 08-670-13C

  • / Each
 

Description & Specifications

Specifications

Product Type Spectra/Por™ 6 Dialysis Tubing
Length (Metric) 10m
Temperature Range (Metric) 4° to 121°C
Temperature Range (English) 39.2° to 249.8°F
Diameter (English) 0.94 in.
Length (English) 33 ft.
MWCO 2 kd
Diameter (Metric) 24mm
Quantity Each
Formulation Packaged wet in 0.05% sodium azide
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Material Regenerated Cellulose

This ready-to-use membrane is packaged wet in a preservative solution of 0.05% sodium azide. Simply rinse away the preservative solution with DI water, load the sample and dialyze.

  • Pre-wetted & ready-to-use, no glycerin removal required
  • 8 MWCO's ranging from 1 kD - 50 kD
  • Good chemical compatibility with dilute strong acids and bases, conc. weak acids & bases, many alcohols and some organics
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • Ideal for sample prep, desalting, buffer exchange, pH change, protein purification, etc.
  • Economic 10 meters/pkg

Do not tie knots in tubing; use closures

Accessories

Accessories for Catalog Number: 0867013C

Item Mfr. No. Item Description Price Per Unit Quantity  
08-750-24C
Spectrum™ Dialysis Tubing Closures: Spectra/Por™ Standard Type
spectrum™
SML132737
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal Pack of 10 Each for $295.26