Please login to your online account to display your discounted pricing

Spectrum™ Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets


Clear, flexible and sturdy Standard RC (regenerated cellulose) in single layer square sheets that can be used with a variety of dialysis equipment or can be cut into specific dimensions for attaching to support frames for customized diagnostic assays, equilibrium studies, desalting, protein purification, etc.

Manufacturer: spectrum™  SML132712

 View more versions of this product

Catalog No. 08-67121

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Dimensions (L x W) 5.9 x 5.9 in. (150 x 150mm)
Operating Temperature (English) 39° to 250°F
Color Clear
Operating Temperature (Metric) 4° to 121°C
Product Type Spectra/Por™ 4 Dialysis Membrane Flat Sheets
Length (Metric) 150mm
Width (English) 5.9 in.
MWCO 12 to 14 kd
Length (English) 5.9 in.
Membrane Type Spectra/Por™ 4
Width (Metric) 150mm
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Quantity 25 Sheets