Please login to your online account to display your discounted pricing

Spectrum™ Spectra Mesh™ Woven, Nylon Filters


Manufacturer: spectrum™  SML145908

 View more versions of this product

Catalog No. 08-671-92

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Thickness 78μm
Material Nylon
Format Circle
Diameter (Metric) 90mm
Particle Retention 100μm
For Use With (Application) Hazardous waste, drinking water purification, Acid rain measurement, suspended soils, air/asbestos, coal dust analysis, oceanography, trace element analysis