Please login to your online account to display your discounted pricing
Spectrum™ Refrig/Arrangers™ Refrigerator Storage Trays
The ideal way to conserve valuable space in refrigerators and freezers (-90°C) as well as organize the laboratory
$183.30 - $183.30
Specifications
Material Polystyrene
Type Refrigerator or Arranger Tray
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
14-334-18
spectrum™
SML183550
Medium Each for $183.30
14-334-19A
spectrum™
SML183554
Large Each for $183.30
14-334-19B
spectrum™
SML183556
Small Each for $183.30
Description & Specifications
Specifications
Material Polystyrene
Type Refrigerator or Arranger Tray
Made of high-impact polystyrene, these lightweight, sturdy Storage Trays are perfect for safe carrying and storage of glass vials or small bottles.
  • Available in channel and grid designs with a raised front edge designed for labeling and preventing spills
  • Efficient Use of Valuable Space
  • Protects Glass and Plastic Containers
  • Transport from Bench-top to Fridge
  • Secondary Containment for Spills
  • Freezer Storage as low as -90 C
  • 3 Channel Tray Sizes
  • 2 Grid Tray Size
    • Available in channel or grid (individual compartments) configuration. Overall, 13 1/2L × 16W × 2 1/4 in.H (34.3 × 40.6 × 5.7cm).