Spectrum™ Drawer/Ganizer™ Portable Organizer Trays
Rigid 1/8 in. polystyrene trays with easy-gripping folded edges for transporting lab items and mobilizing experimental activities
$79.83 - $91.41
Specifications
Material Polystyrene
Thickness 0.32cm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Size Type Price Quantity  
Catalog Number Mfr. No. Includes Size Type Price Quantity  
08-607-052 Spectrum™
SML155585
4 Narrow 42 x 7cm compartments Small Pipette and Column Organizer Tray Each for $79.83
08-607-051 Spectrum™
SML155510
42 x 7cm 6 Narrow compartments Large Pipette and Column Organizer Tray Each for $86.48
08-607-053 Spectrum™
SML155495
16 Square 11 x 10cm compartments Large Stopper and Fitting Organizer Tray Each for $86.48
08-607-054 Spectrum™
SML155570
12 Square 10 x 10cm compartments Small Stopper and Fitting Organizer Tray Each for $79.83
08-607-055 Spectrum™
SML155450
15 Various size compartments Large Parts and Chromatography Tray Each for $86.48
08-607-056 Spectrum™
SML155645
7 Various size compartments Small Parts and Chromatography Organizer Tray Each for $84.38
08-607-061 Spectrum™
SML155465
4 Equal 23 x 20cm compartments Small Utility and Equipment Organizer Tray Each for $86.48
08-607-060 Spectrum™
SML155600
5 Various size compartments Small Clamp and Instrument Organizer Tray Each for $84.38
08-607-059 Spectrum™
SML155525
7 Various size compartments Large Clamp and Instrument Organizer Tray Each for $86.48
08-607-058 Spectrum™
SML155675
Five 27 x 6cm and two 16 x 14cm compartment Small Micropipette Organizer Tray Each for $79.83
08-607-057 Spectrum™
SML155430
Seven 27 x 6cm and two 23 x 23 compartments Large Micropipette Organizer Tray Each for $91.41
08-607-062 Spectrum™
SML155630
4 Equal 20 x 15cm compartments Small Utility and Equipment Organizer Tray Each for $79.83
Specifications
Description
Deep compartments are ideal for holding large quantities or providing secondary containment for spills. Also designed to fit lab drawers, refrigerators and bench tops to protect supplies, organize valuable space and promote laboratory safety.
  • Designed for portability and storage in drawers
  • Protects supplies, promotes safety and provides secondary containment
  • Six tray configurations available in two sizes (large and small)
Description & Specifications
Specifications
Polystyrene
0.32cm