Please login to your online account to display your discounted pricing

Spectrum™ Drawer/Ganizer™ Portable Organizer Trays


Rigid 1/8 in. polystyrene trays with easy-gripping folded edges for transporting lab items and mobilizing experimental activities

Manufacturer: spectrum™  SML155495

 View more versions of this product

Catalog No. 08-607-053

  • / Each
 

Description & Specifications

Specifications

Thickness 0.32cm
Size Large
Includes 16 Square 11 x 10cm compartments
Type Stopper and Fitting Organizer Tray
Material Polystyrene