Please login to your online account to display your discounted pricing
Spectrum™ Drawer/Ganizer™ Cylinder Spacers
Sturdy polystyrene racks that slip over a cabinet door edge (1.9 cm thickness) or mounts directly to the wall to provide safe, inverted storage and organization of lab cylinders, ideal for drying glass ware
$291.00 - $306.48
Products
Catalog Number Mfr. No. Price Quantity    
14-334-17B
spectrum™
SML130045
Pack of 2 Each for $306.48
14-334-17A
spectrum™
SML130030
Pack of 2 Each for $291.00
Description & Specifications
Two sizes available:
  • Small Cylinder Spacer holds 4 smaller cylinders (25mL - 100mL)
  • Large Cylinder Spacer holds 3 larger cylinders (250mL - 1L)