Spectrum™ DynaGard™ Syringe Filter Tips
With looped hollow fiber membrane to maximize surface area and reduce fouling
$413.43 - $708.11
Specifications
Inlet Style Female Luer Lok
Outlet Style Male Luer Taper
Pore Size 0.2μm
Type Syringe Filter Tips
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Material Sterile Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Material Sterile Volume (Metric) Price Quantity  
08-671-8B Spectrum™
DG2M110200
Mixed Cellulose Esters (MCE) No <5mL Pack of 200 Each for $708.11
08-671-8F Spectrum™
DG2M330100
Mixed Cellulose Esters (MCE) No 5 to 20mL Pack of 100 Each for $413.43
Specifications
Description
  • Narrow housing design minimizes hold-up volume and enables easy aspiration and dispensing in ampules, test tubes and other small vessels
  • Available with either 0.2μm hydrophilic mixed cellulose ester (ME) fibers for use with aqueous solutions or 0.2μm hydrophobic polypropylene (PP) fibers for use with organic solvents and alcohols
Description & Specifications
Specifications
Female Luer Lok
Male Luer Taper
0.2μm
Syringe Filter Tips