Please login to your online account to display your discounted pricing
Spectrum Chromatography™ Tubing
$131.57 - $251.77
Specifications
For Use With (Application) Chromatography
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Length (Metric) Material Price Quantity    
11-189-24
spectrum chromatography
SML123746
0.0625 in. 0.1875 in. 3m Silicone Each for $131.57
11-189-25
spectrum chromatography
SML123748
0.0625 in. 0.1875 in. 15m Silicone Each for $251.77
Description & Specifications
Specifications
For Use With (Application) Chromatography
  • Softest and most flexible tubing
  • Translucent white
  • Easily locate meniscus
  • Pure polymerized silicones with nothing to leach out