Spectrum™ Spectra/Chrom™ Aqueous Chromatography Columns
$1,176.04 - $1,176.04
Specifications
For Use With Aqueous solutions including most Acids, bases, and Salts
Material Fluoropolymer Resin, Polypropylene, Borosilicate Glass, and Nitrile Rubber
Type Spectra/Chrom™ Aqueous chromatography column
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Price Quantity  
06-651-5C View Documents Spectrum™
124040*
50mm 90cm Each for $1,176.04
Specifications
Specifications
Aqueous solutions including most Acids, bases, and Salts
Fluoropolymer Resin, Polypropylene, Borosilicate Glass, and Nitrile Rubber
Spectra/Chrom™ Aqueous chromatography column