Spectrum™ Aqueous Chromatography Columns
Manufacturer:  Spectrum™ SML124014
 View more versions of this product
Catalog No. 06-651-2D
Description
Description
Specifications
Specifications
Standard Buffers for high quality, classic chromatographic separation
Aqueous Chromatography Column
106 mL
Borosilicate Glass
15 mm
45 psi
600 mm
0.18 in. I.D. Tubing connectors